ContactUsLinkComponent
GC Multiresidue Pesticide Kit, Restek
GC Multiresidue Pesticide Kit, Restek
# de catalogue 75835-044
Fournisseur:  Restek
Numéro CAS:  
GC Multiresidue Pesticide Kit, Restek
# de catalogue 75835-044
Fournisseur:  Restek
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    GC Multiresidue Pesticide Kit
  • Size:
    Contains 1 ml each of these mixtures
  • Cat. no.:
    75835-044
  • Supplier No.:
    32562
Spécifications
A propos de cet article
Accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
  • Formulated and grouped for maximum long-term stability
  • Quantitatively tested to confirm composition
  • Contains 1 ml each of these mixtures
GC multiresidue pesticide kit consist of GC multiresidue pesticide standard #1, GC multiresidue pesticide standard #2, GC multiresidue pesticide standard #3.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements. Comprehensive 203-compound kit covers food safety lists by the FDA, USDA.
Ordering information: * NOTE: When combining a large number of compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.
Customers Who Bought This Also Bought