ContactUsLinkComponent
QuEChERS Internal Standard Mix for GC-MS Analysis (6 Components), Restek
QuEChERS Internal Standard Mix for GC-MS Analysis (6 Components), Restek
Catalog # 75835-214
Supplier:  Restek
CAS Number:  
QuEChERS Internal Standard Mix for GC-MS Analysis (6 Components), Restek
Catalog # 75835-214
Supplier:  Restek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Concentration:
    In acetonitrile 5 ml/ampule
  • Description:
    QuEChERS Internal Standard Mix for GC/MS Analysis
  • Size:
    5 ml/Ampule
  • Cat. no.:
    75835-214
  • Supplier No.:
    33267
Specifications
About this item
Ready to use for QuEChERS extractions—no dilutions necessary.
It consits of PCB 18 (50 μg/ml), PCB 28 (50 μg/ml), PCB 52 (50 μg/ml), triphenylmethane (10 μg/ml), triphenylphosphate (20 μg/ml), tris-(1,3-dichloroisopropyl)phosphate (50 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Customers Who Bought This Also Bought