SP Wilmad-LabGlass Polished Glass Stirrer Shafts, SP Industries
Supplier: SP Industries
Certificates
About this item
Precision ground and polished, these borosilicate glass stirrer shafts have a 10mm O.D
For use with LG-9475T bearing with PTFE bearing surface, or other bearings with a PTFE interface with the same bore. Features a side button at bottom for attachment of any of the stirrer/stirring blades (LG-9505 through LG-9511T).
Customers Who Bought This Also Bought