VWR® Bead Mill Homogenizer, 120 V | Avantor
VWR® Bead Mill Homogenizer, 120 V
Bead Mill Homogenizer, 25 µl-5 ml
Catalog # 75840-022
Product available on GSA Advantage®
Supplier:   VWR International
VWR® Bead Mill Homogenizer, 120 V
Catalog # 75840-022
Supplier:  VWR International
Product available on GSA Advantage®
VWR® Bead Mill Homogenizer, 120 V
VWR® Bead Mill Homogenizer, 120 V
Bead Mill Homogenizer, 25 µl-5 ml
Price
$14,428.05
Log in pour la disponibilité
Each
Product Details & Documents
The VWR® Bead Mill is part of the newest generation of Bead Mill homogenizers. This economical system is specifically designed for grinding, lysing and homogenization of up to 16 samples. Using sample tubes pre-filled with a variety of lysing beads, the VWR® Bead Mill vigorously and uniformly shakes the tubes providing an efficient, consistent, high-yield and quality homogenization usually in less than 40 seconds.
  • 16 sample capacity
  • Run time 1 second to 10 minutes
  • 0.8 to 6 m/s
  • Up to 99 programs
  • Compatible with VWR® pre-filled bead tubes
  • TUV Certified
  • Two-year warranty
The VWR® Bead Mill simultaneously homogenizes 12×0.5, 4×1.5, 12×2, or 4×7ml samples.
The VWR® Bead Mill is an ideal solution for releasing DNA, RNA and proteins from tough samples.
It does not require a cool down between runs which allows for non-stop use and high throughput. The unit features a convenient front loading design with snap in tube carriage.
Certifications: c TUV US Certified
Ordering information: Includes the Bead Mill Drive Unit, Tube Carriage, User Manual, and a two-year warranty. Bead Mill disposable bead tubes sold seperately.
Spécifications
  • Désignation:
    Bead Mill Homogenizer
  • Sample Size:
    25 µl-5 ml
  • Cat. No.:
    75840-022
  • Run time:
    1 second to 10 minutes
  • Speed range:
    0.8 to 6 m/s
  • No. of cycles:
    1
  • Dimensions:
    30.5×43.2×36.8 cm
  • Electrical:
    120 VAC
  • Weight:
    24 kg (45 lbs.)