EZwaste™ Safety Tray Secondary Containers, Foxx Life Sciences
Supplier: VWR International
Certificates
About this item
An ideal solution for preventing leakage and spillage during carboy storage or transport.
- Tough and durable
- Designed with the capacity to hold 115% of carboy content
EZwaste™ safety tray secondary containers are a perfect solution for preventing leakage and spillage from EZwaste™ solvent waste system containers.
Customers Who Bought This Also Bought