Safety Disposal Cans, Galvanized Steel, Eagle Manufacturing
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
These galvanized steel disposable cans have a red powder coat finish
- Spout cap opens to a wide 80° angle
- Equipped with a frame arrestor
Can be locked open for conveniently emptying various type containers. Cap assembly opens automatically to relieve excessive internal vapor pressures.
They provide an intermediate station for temporary storage of liquid wastes.
Certifications: FM approved.
Customers Who Bought This Also Bought