Graduated Cylinder Support, Accurate Wirecraft
# de catalogue 24795-003
Fournisseur: Accurate Wirecraft
Spécifications
- Description:Graduated Cylinder Support
- Height:43 cm (17")
- Cat. no.:24795-003
- Supplier No.:252018BK
Spécifications
A propos de cet article
Holds up to 25 cylinders and revolves for removal of dry cylinders.
- For drying and storing graduated cylinders
- 51 cm diameter drain pan made of rust proof aluminum
Made of strong steel rods electrically spot-welded, coated with cushioning black vinyl.
Customers Who Bought This Also Bought