SP Wilmad-LabGlass PTFE Thermometer Adapter with Compression Cap, SP Industries
# de catalogue 89087-526
Fournisseur: SP Industries
Spécifications
- Fits:[ST] 10/18 and [ST] 10/30 outer joints
- Joint Size:[ST] 10/18
- Supplier No.:LG-1453-100
Spécifications
A propos de cet article
This inner joint thermometer adapter has a threaded compression cap, four saddle O-Rings, and one Viton® O-Ring for use with plain stem thermometers and inlet tubes.
- Threaded compression cap, four saddle O-Rings, and one Viton® O-Ring included
- Accommodates thermometers and inlet tubes
- Made of PTFE
The adapter accepts thermometers with outer diameters from 3 to 7mm.
Customers Who Bought This Also Bought