Laboratory Ball Valve, WaterSaver Faucet
# de catalogue VFL4200-158WS
Fournisseur: WaterSaver Faucet
Spécifications
- Description:Laboratory Ball Valves
- Cat. no.:VFL4200-158WS
- Supplier No.:L4200-158WS
Spécifications
A propos de cet article
For installation on vertical or horizontal surfaces (such as pipe chases and service carriers) where there is access behind surface during installation.
- Forged brass valve body
- Forged brass lever handle with color-coded index disc
Inlet furnished with 3/8" IPS mounting shank (assembled), locknut and washer. 3/8" NPT male inlet. 3/8" NPT female outlet with removable ten serration hose end.
Customers Who Bought This Also Bought