Pressure Regulator Fixtures, WaterSaver Faucet
# de catalogue 97014-324
Fournisseur: WaterSaver Faucet
Spécifications
- Description:Pressure Regulator
- Cat. no.:97014-324
- Inlet Pressure:5–300 psi
- Outlet Pressure:5 - 125 psig
- Supplier No.:L2873-771WSA
Spécifications
A propos de cet article
Pressure regulator used for compressed air and inert high purity gases, including nitrogen, helium, argon and oxygen.
- 3/8" NPT male inlet
- Removable quick connect fitting with 1/4" NPT male plug
- Non-relieving type with brass body and neoprene diaphragm
Fine control needle valve for precise flow control. Valve has forged brass body, floating tapered stainless steel needle and replaceable stainless steel seat.
Customers Who Bought This Also Bought