Lovibond® MD 100 COD for Photometer, Tintometer
# de catalogue 10752-412
Fournisseur: Lovibond
Small Business Enterprise
Numéro CAS:
Spécifications
- Description:MD 100 COD Photometer
- Model:MD 100
- Battery life:17 h or 5000 tests
- Wavelength accuracy:±1 nm
- Operating humidity:30 - 90% RH
- Accuracy:3% FS (T=20...25°C)
- Display:LCD, backlit
- Interfaces:Infrared interface for test data transfer
- Keypad:4 button membrane keypad
- Operating temperature:5…40 °C
- Optics:LED, interference filters (IF) and photo sensor in transparent sample chamber
- Power supply:4× AAA batteries
- Storage capacity:16 data sets
- Wavelengths:430, 530, 560, 580, 610, 660 nm
- Weight:~0,26 kg
- W×D×H:155×75×35 mm
- Cat. no.:10752-412
- Range:3 - 150 mg/L, 15 - 300 mg/, 20 - 1500 mg/, 200 - 15000 mg/ O₂
- Supplier No.:276120
Spécifications
A propos de cet article
The MD 100 COD Photometer provides photometric detection to measure the COD concentration from 0 to 15000 mg/l O2 and a simple button layout, robust waterproof design and large backlit display make the MD100 easy to use and ideal for testing in the field, laboratory or plant room.
- Accurate (3% FS), reproducible results using high quality interference filters with long-life LEDs
- Internal ring memory for 16 data sets
- Real time clock and date
- Optional infrared interface for transfer of test data to PC or printer
The unit supplies accurate, reproduceable results using high quality interference filters with long-life LEDs as a light source. Tests are conducted using the standard small scale sealed tube method. Lovibond® Vario COD vials are programmed for measurements of 0-150, 0-1500 or 0-15000 mg/l.
Delivery: Instrument supplied with carry case, 4 x AAA batteries, 3 x round glass vials with lids, plastic syringe (1 and 2 mL), COD vial adapter, guarantee sheet, certificate (COC), and instruction manual.
Customers Who Bought This Also Bought