Optimus™ Pedestal Mounted Eye/Face Wash System, Speakman®
Fournisseur: Speakman
Certificats
A propos de cet article
Pedestal mounted eye/face wash system equipped with a dual aerated eyewash to deliver soothing relief.
- Two individual face pads producing 66 individual, full-coverage sprays
- Elevated drain allows for easy recommended testing
- Three year limited warranty
Steel pedestal with a DuraJade powder-coated or stainless steel finish provides a solid, versatile mount.
Certifications: ANSI Z358.1 certified and OSHA compliant.
Delivery: Pedestal mounted eye and face wash system includes a 1¹/₄" diameter steel pedestal.
Customers Who Bought This Also Bought