Optimus™ Wall Mounted Eye/Face Wash System, Speakman®
Supplier: Speakman
Certificates
About this item
Wall mounted eye/face wash system equipped with a dual aerated eyewash to deliver soothing relief.
- Two individual face pads producing 66 individual, full-coverage sprays
- Three year limited warranty
A simple push of the intuitive handle activator to instantly deliver soaking, soothing streams onto all affected areas. Equipped standard with an extended mounting arm and the capability to be installed at any height.
Certifications: ANSI/ISEA Z358.1 certified and OSHA compliant. Meets ADA specifications.
Customers Who Bought This Also Bought