Optimus™ Combination Emergency Shower and Eye/Face Wash System, Speakman®
Fournisseur: Speakman
Certificats
A propos de cet article
Combination emergency shower and eye/face wash system equipped with a dual aerated emergency eye wash to deliver soothing relief.
- Combined with emergency shower with 8" Ø shower head
- Two individual face pads producing 66 individual, full-coverage sprays
- Three year limited warranty
Features a highly visible yellow ABS plastic or corrosion resistant stainless steel bowl attached to a powder-coated steel emergency shower.
Certifications: ANSI/ISEA Z358.1 certified, OSHA compliant. Pipe fittings can be customized to meet ADA height specifications.
Customers Who Bought This Also Bought