Vertical Rods, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
Certificats
A propos de cet article
Vertical rods are machined with the top end rounded and the bottom end tapered to fit in to a rod socket.
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
48,82 $ to 442,63 $