PEEK Ferrules, MicroSolv
# de catalogue 97055-862
Fournisseur: MicroSolv Technology Corp
Spécifications
- Description:Double Cone Ferrule
- Material:PEEK
- Ferrule Size:¹⁄₁₆"
- Cat. no.:97055-862
- Supplier No.:47320-35
Spécifications
A propos de cet article
Double cone ferrules for use with 2 piece male nuts.
Customers Who Bought This Also Bought