SP Wilmad-LabGlass Four-Neck Flat Flange Reaction Vessel Covers, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
Featuring a center neck and three side necks with full-length outer Standard Taper joints, these borosilicate glass four-neck reaction vessel lids/covers have a flat flange
NOTE: LG-8087 is designed to be used with LG-8080, LG-8084, LG-8085, and LG-8089 cylindrical reaction vessels. LG-8087A is designed to be used with LG-8082, LG-8084, LG-8085, LG-8089 vessels.
Use LG-7316 clamps.
Customers Who Bought This Also Bought