SP Wilmad-LabGlass Three-Neck Flat Flange Reaction Vessel Covers, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
Featuring a center neck and two side necks with full-length outer Standard Taper joints, these borosilicate glass three-neck reaction vessel lids/covers have a flat flange with 53/8" O.D
Designed to be used with LG-8080 cylindrical reaction vessels. All sizes use LG-7316-106 clamps.
Customers Who Bought This Also Bought