Kestrel DROP D2 Wireless Temperature and Humidity Logger
# de catalogue 470222-125
Fournisseur: NIELSEN-KELLERMAN CO
Spécifications
- Description:Kestrel Drop D2 temperature and humidity monitor
- Cat. no.:470222-125
- W×D×H:1×1.8×2.4"
- Supplier No.:0720RED
Spécifications
A propos de cet article
The easiest way to track environmental data
- View data wirelessly via Bluetooth® low energy on your phone or tablet with the free Kestrel LiNK app
- Store thousands of data points and get months of logging on a single coin-cell battery - customize logging rate, connections and alerts
A Kestrel DROP is the world’s easiest-to-use rugged environmental data logger. The Drop D2 allow you to monitor and record the conditions of any environment and easily access and chart real-time and historical data on your phone, tablet or other smart device. The LiNK app is available for both iOS and Android. Measures: Dew Point, Heat Stress Index, Relative Humidity, Temperature.
Ordering information: App requirements include iOS 6 installed iPhone 4s, iPad 3rd Generation, iPod Touch 5th Generation and above. Android 4.3 and higher. Works with most Android devices with Bluetooth® low energy, including Samsung Galaxy, Nexus 4, Motorola Droid and other recent generation phones and tablets.
Customers Who Bought This Also Bought