Kestrel 3500 Fire Weather Meter
# de catalogue 470222-121
Fournisseur: NIELSEN-KELLERMAN CO
Spécifications
- Description:Kestrel 3500 Fire weather meter
- Cat. no.:470222-121
- W×D×H:4.8×1.7×0.7"
- Supplier No.:0835FWORA
Spécifications
A propos de cet article
The Kestrel 3500 has the most functions of any Kestrel Meter in the 3000 series. It measures wind speed, relative humidity, temperature, wet bulb temperature (psychrometric), altitude, pressure trend, barometric pressure, heat stress index, dew point temperature, wind chill.
- Easy to use system scrolls through 9 different environmental conditions
- Waterproof and floats
- Large easy to read display
Slip on protective cover included.
Customers Who Bought This Also Bought