Kestrel DROP D3 Fire Weather Monitor
# de catalogue 470222-119
Fournisseur: NIELSEN-KELLERMAN CO
Spécifications
- Description:Kestrel D3 Fire weather meter
- Cat. no.:470222-119
- W×D×H:2.4×1.8×0.9"
- Supplier No.:0730FWORA
Spécifications
A propos de cet article
The drop can be easily placed in the field and begin collecting environmental data.
- View, chart, record and monitor your environmental conditions
- Real time, Wireless, Graphing functionality
- Access real time or chart the data via Bluetooth Low Energy (BLE) on your smart device
- Log options: 2s, 5s, 10s, 20s, 30s, 1min, 2min, 5min, 10min, 20min, 30min, 1hr, 2hr, 6hr, 12hr
DROPs are so small they can easily be clipped to a pack or belt loop, and so affordable they can be placed in multiple locations to provide unmanned monitoring. The DROP is powered by LiNK wireless data connectivity and the LiNK iOS and Android App – allowing for remote view of DROP readings as well as hands-free automatic data upload for review whenever the opportunity presents. Measures: Temperature, Relative Humidity, Heat Stress Index, Dew Point Temperature, Density altitude, Station pressure
Ordering information: App requirements include iOS 6 installed iPhone 4s, iPad 3rd Generation, iPod Touch 5th Generation and above. Android 4.3 and higher. Works with most Android devices with Bluetooth® low energy, including Samsung Galaxy, Nexus 4, Motorola Droid and other recent generation phones and tablets.
Customers Who Bought This Also Bought