SteriPlast® Spoons for Foodstuffs, PS, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
The SteriPlast® spoon ideal for food, animal feed and pharmaceutical industries.
- Can be used as part of HACCP/IFS foreign object management
- Individually packaged and sterilized by gamma rays
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
Certifications: Production, assembly and packaging according to cleanroom class 7. Complies with FDA regulations.
Ordering information: Also available with long handle (11.8") and curved bowl in different sizes - 5 ml (0,17 oz.), 10 ml (0.34 oz.) or 20 ml (0.68 oz.).
Customers Who Bought This Also Bought