VWR® Spatulas, PS
Catalog # 10820-353
Supplier: VWR International
Specifications
- Color:White
- Description:Spatula
- Length:26.3 cm (10.3")
- Cat. no.:10820-353
Specifications
About this item
Spatula with its long, ergonomic solid handle and pointed blade can be easily pierced into paper and plastic bags.
- Ideal for the sampling of powder, granules and pastes
- Individually packed, gamma radiation sterilized
Certifications: Production, assembling and packaging according to cleanroom Class 7 (10,000). Complies with food and FDA guidelines.
Customers Who Bought This Also Bought