Gas-Tight Stainless Steel Drum Pumps, Burkle
Supplier: Avantor
Certificates
About this item
Gas-tight barrel pump with flexible discharge hose for filling off into large canisters and containers.
- For bottle openings from 16 - 49 mm
- Immersion tube Ø 32 mm
- Gas-tight barrel screw joints with 2" fine thread
- The pump can be hermetically sealed with two ball valves
Convenient transfer because the canister can stay on the floor. The connection cone is held onto the container. Parts in contact with media - stainless steel (1.4301), PTFE, nickel-plated brass, PA that can conduct electricity.
Customers Who Bought This Also Bought