Laboratory Printers, METTLER TOLEDO®
Fournisseur: Mettler Toledo
Certificats
A propos de cet article
These laboratory printers are compatible with METTLER TOLEDO Lab Instruments.
- Eliminate transcription errors, speed up working processes and ensure traceability
- Switch between paper, continuous self-adhesive labels and labels
- Just plug in and start printing
- Includes strip printing unit (24 characters/line)
Model P-52RUE dot matrix printer supports printing on paper and continuous label and can be connected via RS232, USB and Ethernet interfaces.
The robust model P-56RUE and P-58RUE universal printers offer a built-in real-time clock, and multitude of printing possibilities to help you to eliminate transcription error, speed up your working processes and ensure traceability.
Model P-58RUE printer can also perform statistics, totaling, formulation, and barcode generation.
Ordering information: For compatibility with specific METTLER TOLEDO instruments, please contact your Avantor representative.
Spécifications
- Catalog No:
- CA10147-660
- CA10147-788
- 75982-118
- Interfaces:
- RS232; USB; Ethernet
- RS232; USB; Ethernet
- RS232; USB; Ethernet