Inkjet Microscope Slides, Springside Scientific
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Certificats
A propos de cet article
Inkjet Microscope Slides are manufactured from 3/4 white glass
All slides are standard size, with frosting optimized for Leica inkjet labelers, ensuring good barcode reads with each use. Slides are available charged or uncharged.
Ordering information: Slides are packed 72 per cardboard box. Each box is individually sealed. Each case contains 20 boxes.
Customers Who Bought This Also Bought