Adhesive/Charged Microscope Slides, Springside Scientific
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Certificats
A propos de cet article
Microscope slides are manufactured from 3/4 white glass and feature a positive charge for improved adhesion
All slides are standard size, with frosting optimized for hot foil tape labelers such as the Slidemate and Primera.
Ordering information: Slides are packed 72 per white cardboard box. Each box is individually sealed. Each case contains 20 boxes.
Customers Who Bought This Also Bought