A propos de cet article
Recommended for use in the cultivation of a wide variety of fastidious and nonfastidious microorganisms.
- For fastidious and nonfastidious microorganisms
- With neutralizing additives
Lecithin & Tween® 80 help in neutralizing residual sanitizing chemicals and help in emulsifying samples. This product is not intended for use in diagnosing human disease.