Orum International Trio.Bas™ Satellite Kit for ISOLATOR/RABS Units
Fournisseur: Hardy Diagnostics
Certificats
A propos de cet article
A remote aspitating head satellite, available with contact plate or petri plate format, with 5 m long cable, for use with the RABS/ISOLATOR microbial air sampling instrument.
- AISI 316 rated stainless steel connects to RABS/ISOLATOR command unit (sold separately)
The TRIO.BAS™ instruments utilize the impaction method of air sampling. The air, passing through an aspirating head, impacts onto an appropriate agar plate that collects bacteria or other organic substances possibly present in the sampled area.
Per USP 797, 'Impaction is the preferred method of active air sampling. Use of settling plates for qualitative air sampling cannot be relied upon and shall not be used solely to determine the quality of air in the controlled environment.'
Delivery: TRIO.BAS™ Satellite kit contains: (1) Satellite air sampling aspirating chamber, (1) stainless steel aspirating cover, 5 m attachment cable and light carrying case.
Spécifications
- Catalog No:
- 75838-738
- 75838-736
- Flow rate:
Spécifications
Informations connexes