ContactUsLinkComponent
Oxoid PALCAM Selective Supplement, Hardy Diagnostics
Oxoid PALCAM Selective Supplement, Hardy Diagnostics
# de catalogue CA10790-400
Fournisseur:  HARDY DIAGNOSTICS CA
Numéro CAS:  
Supplement
Oxoid PALCAM Selective Supplement, Hardy Diagnostics
# de catalogue CA10790-400
Fournisseur:  HARDY DIAGNOSTICS CA
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    PALCAM Selective Supplement
  • Cat. no.:
    CA10790-400
  • Supplier No.:
    SR150E
Spécifications
A propos de cet article
A selective supplement or the isolation of Listeria monocytogenes.
A selective supplement for the isolation of Listeria monocytogenes when used with PALCAM Agar Base CM877B. Vial contents (each vial is sufficient for 500ml of medium): Polymixin B 5mg Acriflavine hydrochloride 2.5mg Ceftazidime 10mg 10mg
Customers Who Bought This Also Bought