Spécifications
- Description:CampyGen Compact
- Size:20 Sachets
- Cat. no.:CA10790-378
- Supplier No.:CN020C
Spécifications
A propos de cet article
Simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.
The Oxoid CampyGen and CampyGen Compact are simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.
Like the AnaeroGen, the CampyGen also requires no catalyst and no water activation steps. No dangerous gas is produced. The final atmosphere is reduced to 6% oxygen, and supplemented with 14% carbon dioxide which is ideal for growth of Campylobacter and other microaerophilic organisms. Sachet, bags and clips are sold separately.