Ultratorch® Solder Irons, Master Appliance
Fournisseur: Master Appliance
Certificats
A propos de cet article
The versatility in size and power of these Microtorch® butane torches offer something for everyone, and no matter what job you have on your “to do” list, we have the portable soldering iron and butane torches perfect for any task at hand.
- Professional quality
- Self-igniting
- Butane powered soldering/heat tool and torch
- Oxygen free, copper based Ultra tip soldering tips
- Thick iron and chrome plating for long life and high thermal conductivity
- Completely portable
- Ready to use in 30-seconds
- Adjustable temperature control
- Patented gas catalyst reaction system
Soldering temperature up to 932-Degree Fahrenheit, ideal for use with today's lead free solder
Use to pinpoint heat, remove surface mount components, repair vinyl, heat shrink tubing, solder and desolder, spot dry and more.
Sturdy metal tank with fuel viewing window holds enough fuel for 2 hours of operation. The soldering tips are "tinned" at the tip of the tip with lead-free solder thereby will have slight color variance from the rest of the tip.
Spécifications
- Catalog No:
- 94025-818
- 94025-824
- Length:
- 260 mm (10.3")
- 208 mm (8.3")