Military Switchboard Anti-Fatigue Matting, Wearwell®
Catalogue # 89021-400
Fournisseur: Wearwell
Spécifications
- Color:Black
- Description:Smooth Military Switchboard Matting, Full Roll
- Length:2286 cm (75 ft.)
- Thickness:3.2 mm (⅛")
- Cat. no.:89021-400
- Width:91 cm (3 ft.)
- Supplier No.:711.18X3X75BK
Spécifications
À propos de cet article
Military Switchboard matting prevents life-threatening electrical shocks in high-voltage environments.
- Meets Mil Spec 15562-G, Type II, Class 3
- Dielectric Strength: 30,000 volts
- Easy to clean Smooth (Type II) surface
- Exceptional ozone and oil resistance
This non-conductive matting delivers a dielectric strength of 30,000 volts. The easy to clean surface offers exceptional ozone and oil resistance. Ideally suited for use in dry environments.
Certifications: Meets ASTM D1056. –20 to 160°F. Passes Mil. 15562-G and FMVSS 302.