ErgoDeck Heavy Duty with GRITSHIELD™ Modular Anti-Fatigue Flooring, Wearwell®
Supplier: Wearwell
Certificates
About this item
Heavy duty compound allows carts to roll easily, while the box-spring design provides structure.
- Standing relief in wet or dry applications
- Customize in 3" increments to fit any area or configuration
- Positive Interlocking System prevents separation
- Easily replace single pieces or sections as needed to reduce costs
- 100% silicone-free PVC tiles provide maximum durability
A comfortable, ergonomic floor that will withstand heavy use. The patented LockSafe® Positive Interlocking System delivers secure connections. Perfect for lean manufacturing, tiles can be unlocked and repositioned as work centers change to fit customer needs. The optional 6" side safety ramps and corners have countersunk holes for semi-permanent installation. Available in a solid and drainage version, ErgoDeck Heavy-Duty with GRITSHIELD™ delivers 40% more traction and withstands heavy use and cart traffic.
Certifications: Meets ASTM F1677-05. Dry: COF = 1.1/Wet: COF = 0.93.
Customers Who Bought This Also Bought