Abrasive Coated Kushion Walk Industrial Runner, Wearwell®
Fournisseur: Wearwell
Certificats
A propos de cet article
Abrasive Kushion Walk features the GRITSHIELD™ surface, designed for slip resistance.
- Industrial tough and abrasion resistant
- Durable textured surface guards against slipping
- Drainage slots allow runoff and aeration
- Highly cut resistant
- Excellent choice for covering grated walkways
Abrasive Coated Kushion Walk features the unique GRITSHIELD™ surface, specifically designed for high COF [Coefficient of Friction] in the slickest locations. With slips and falls being such a dangerous nuisance in today's workplace, Abrasive Coated Kushion Walk offers an effective standing solution in areas requiring high traction.
Certifications: Meets ASTM F1677. Dry: COF = 1.03/Wet: COF = 0.81.
Customers Who Bought This Also Bought