Difco™ Columbia Broth, BD
# de catalogue CA90004-548
Fournisseur: BD
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Difco™ Columbia Broth
- Size:500 g
- Cat. no.:CA90004-548
Spécifications
A propos de cet article
Columbia Broth is used for cultivating fastidious microorganisms.
- Peptones and yeast extract provide nitrogen, carbon, vitamins, and nutrients
- Dextrose provides an additional carbon energy source
- Medium is buffered with Tris
- Cysteine acts as a reducing agent
- Magnesium and iron are added to facilitate growth
Columbia Broth, in the presence of CO2 and supplemented with SPS, is an excellent blood culture medium.
Customers Who Bought This Also Bought