Microtorch® Butane Powered Torches, Master Appliance
Fournisseur: Master Appliance
Certificats
A propos de cet article
Master butane torches are perfect for any task at hand. They are self-contained, butane-powered, heavy-duty, general industrial torches.
- Self-igniting piezo electric ignition that is easy to start and eliminates need for lighter
- Self-contained, metal fuel tank that is rugged and lightweight, easy to use, eliminates bulky canister
- Large fuel capacity, 80 cc self-contained tank for 30-minute continuous run time
- Large sized adjustable blue flame that accommodates many heavy-duty industrial applications with temperatures up to 2500 °F/1300 °C
- Safety lock that prevents unintentional ignition, locks for storage
- Use to braze with special alloy welding rods, heat and bend thin metals, loosen rusted bolts, pop dents out of metal, shrink and terminate heavy wall adhesive lined tubing, solder and desolder, and more
The MT-11 Microtorch Model butane tool from Master Appliance is palm-sized and self-igniting for your convenience.
The GT-70 general industrial torch is a compact, self-igniting butane heat tool made by Master Appliance, and its large blue flame reaches temperatures up to 2500 degrees Fahrenheit. Approved by the CPSC, this model should be refilled with Master Ultratane butane for optimal performance.
Certifications: Meets Consumer Products Safety Commission (CPSC) Requirements
BodyTextAccessories: Refill with Master Ultratane butane for best performance.
Customers Who Bought This Also Bought