Spécifications
- Description:BBL™ CTA Medium™
- Size:500 g
- Cat. no.:CA90000-080
- Supplier No.:211096
Spécifications
A propos de cet article
CTA Medium™ (Cystine Trypticase™ Agar Medium) is a simple, basic medium for the maintenance and detection of motility of a wide variety of microorganisms.
- Cystine and casein peptone provide necessary nutrients
- Carbohydrate fermentation is detected by a visible color change
- Used for the detection of bacterial motility
With added carbohydrates, it is useful for the determination of fermentation reactions.