ContactUsLinkComponent
Difco™ Azide Dextrose Broth, BD
Difco™ Azide Dextrose Broth, BD
# de catalogue CA90003-102
Fournisseur:  BD
Numéro CAS:  
Difco™ Azide Dextrose Broth, BD
# de catalogue CA90003-102
Fournisseur:  BD
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Difco™ Azide Dextrose Broth
  • Size:
    500 g
  • Cat. no.:
    CA90003-102
Spécifications
A propos de cet article
Azide Dextrose Broth is used for cultivating streptococci in water and wastewater.
  • Beef extract and peptones provide essential nutrients
  • Dextrose is a fermentable carbohydrate
  • Sodium chloride maintains osmotic equilibrium
  • Sodium azide inhibits cytochrome oxidase in gram-negative bacteria
Azide Dextrose Broth is specified for use in the presumptive test of water and wastewater for fecal streptococci by the Multiple-Tube Technique.
Customers Who Bought This Also Bought