To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:PIP
- Host:Armenian hamster
- Isotype:IgG
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- Flow cytometry:Yes
- Size:100 µg
- Storage temperature:2-8°C protected from light
- Cat. no.:75834-306
- Supplier No.:65-4888-U100
Spécifications
A propos de cet article
The PIP immunoglobulin is useful as an isotype-matched control. The PIP immunolglobulin is specific for Glutathione S-transferase and can be used as an isotype control for Armenian Hamster IgG antibodies.
Type: Control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: PIP
Conjugation: PerCP-Cy5.5
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: N/A