Continental Fire Assay Crucibles, Anachemia
Fournisseur: VWR International
Certificats
A propos de cet article
Continental Crucibles are specifically designed for precious metal analysis
- For precious metal analysis
- Consistent dimensional tolerance
- Safe and efficient handling
The consistency of their dimensional tolerances and mechanical strength makes manual and automatic handling very safe and efficient. The Continental Crucible is a multi-use crucible, and can be used with most assay tools.
Ordering information: Multiple sizes are available. Contact your Avantor sales Representative for more information.
Customers Who Bought This Also Bought