Hazardous Materials Low-Density Flat Liner, Associated Bag
# de catalogue 89137-984
Fournisseur: Associated Bag
Spécifications
- W×L:38×60"
- Color:Yellow
- Cat. no.:89137-984
- Supplier No.:36-8-100Y
Spécifications
A propos de cet article
Bold colors and printed warnings identify hazardous or contaminated contents.
- Strong bottom seals
- Free of heavy metals
- Yellow
- Printed warnings
Several message options are available.
Weight 13½ lbs.
Packaging: 50 per carton.
Customers Who Bought This Also Bought