Transport Tubes, Starplex Scientific
Supplier: Starplex
Certificates
About this item
Threadless polypropylene round bottom tubes fit easily into racking or automated systems.
- Smooth
- Durable
Tubes are available clear or amber for light sensitive specimens.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.