Environmental Test Chambers, Memmert
Fournisseur: Memmert
Certificats
A propos de cet article
Climatic Test Chamber CTC with Humidity Control
CLIMATIC TEST CHAMBER CTC/TEMPERATURE TEST CHAMBER TTC 100 % AtmoSAFE: In Memmert environmental test chambers CTC and TTC, the perfect atmosphere for climate and temperature tests, specifically in accordance with DIN EN 60068-2-1, 2-2 and 2-3 are simulated. Ramp operation, active humidification and dehumidification of 10 to 98 % and precise temperature control from -42 °C to +190 °C (without humidity) with humidity control from +10 °C to +95 °C provide unlimited flexibility for controlled material and function tests as well as aging tests.
Temperature Test Chamber TTC “Celsius Standard Software. Model size: 256L.
Customers Who Bought This Also Bought