Superwhite Coverglass, Springside Scientific
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Certificats
A propos de cet article
Microscope coverslips are made of pure, super white glass which prevents auto-fluorescence
They work well in all automated coverslippers with no sticking or breaking. Varied sizes are available.
Ordering information: Each plastic box contains 1oz. Each case includes 10 boxes, and is vaccum sealed.
Customers Who Bought This Also Bought