Inkjet Superwhite Microscope Slides, Springside Scientific
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Certificats
A propos de cet article
Inkjet Superwhite Microscope Slides are made of pure, super white glass which prevents auto-fluorescence
They are uniformly 75x25x1mm. Colored frosting is optimized for Leica inkjet labelers, ensuring consistently good barcode reads.
Ordering information: Charged slides are packaged in plastic boxes. Uncharged slides are packaged in ardboard boxes. Each box contains 72 slides, along with fiber desiccant. Each box is individually sealed. Each case contains 20 boxes.
Customers Who Bought This Also Bought