Superwhite Uncharged Microscope Slides, Springside Scientific
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Certificats
A propos de cet article
Microscope slides made of pure, super white glass which prevents auto-fluorescence
All slides are uncharged, and are uniformly 75x25x1mm. Colored frosting is optimized for hot foil tape labelers such as the Slidemate and Primera.
Ordering information: Microscope slides made of pure, super white glass which prevents auto-fluorescence. All slides are uncharged, and are uniformly 75x25x1mm. Colored frosting is optimized for hot foil tape labelers such as the Slidemate and Primera.
Customers Who Bought This Also Bought