Superwhite Adhesive Micro Slides, Springside Scientific
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Certificats
A propos de cet article
Superwhite Adhesive Micro Slides are made of pure, super white glass which prevents auto-fluorescence
Slides are positively charged for adhesion, and are uniformly 75x25x1mm. Colored frosting is optimized for hot foil tape labelers such as the Slidemate and Primera.
Ordering information: Slides are packaged 72 per white plastic box, along with fiber desiccant. Each box is individually sealed. Each case contains 20 boxes.
Customers Who Bought This Also Bought