Omega II Cap Safety Helmet, ERB Safety
# de catalogue 10159-642
Fournisseur: ERB INDUSTRIES, INC.
Spécifications
- Color:White
- Description:Omega II cap safety helmet
- Cat. no.:10159-642
- Supplier No.:19951
Spécifications
A propos de cet article
Omega II cap safety helmets with ratchet knob adjustment.
- 6-point woven nylon ratchet suspension
- Accomodates chin strap
- Certified when headband is installed in reverse position
- Moisture-wicking brow pad is removable, ashable, and replaceable
- Goggle retaining slots on suspension
- Fits head sizes 6.5 - 8
Brow pad is replaceable.
Accessory slots accommodate a wide variety of eye, face, and hearing protection.
Certifications: Meets ANSI/ISEA Z89.1, Type 1, Class C, E and G. SEI ceritified.
BodyTextAccessories: Accessories not inlcuded.
Customers Who Bought This Also Bought