Histology Control Slides, Reticulum, Springside Scientific
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Certificats
A propos de cet article
100% Human Tissue.
Two stained controls in every box. Top-quality, pure white glass slides/coverglass.
Ordering information: Packaged 22 (20 controls plus 2 stained) or 100 (98 controls plus 2 stained) slides in a 25-postion or 100-position plastic slide-box.
Customers Who Bought This Also Bought